4-Amino-5-(2-trifluoromethoxy-phenyl)-4H-[1,2,4]triazole-3-thiol

Lieferant: Apollo Scientific

PC430526-100MG
APOSPC430526-100MGEA 0 EUR
APOSPC430526-100MG
4-Amino-5-(2-trifluoromethoxy-phenyl)-4H-[1,2,4]triazole-3-thiol
4-Amino-5-(2-trifluoromethoxy-phenyl)-4H-[1,2,4]triazole-3-thiol
Formel: C₉H₇F₃N₄OS
Molecular Weight: 276,238 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585448
CAS-Nummer: 1174189-81-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR