Ethyl-4-chlor-6-fluorchinolin-3-carboxylat
Lieferant: Apollo Scientific
Synonyme:
Ethyl 4-chloro-6-fluoroquinoline-3-carboxylate
PC430527-500MG
APOSPC430527-500MGEA
200
EUR
APOSPC430527-500MG
Ethyl-4-chlor-6-fluorchinolin-3-carboxylat
Ethyl-4-chlor-6-fluorchinolin-3-carboxylat
Formel:
C₁₂H₉ClFNO₂ Molecular Weight: 253,66 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173350 CAS-Nummer: 77779-49-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...