2-(4-Fluorophenoxy)-1-ethanamine 95%

Lieferant: Apollo Scientific

Synonyme: 2-(4-Fluorophenoxy)ethylamine

PC430528-500MG
APOSPC430528-500MGEA 203 EUR
APOSPC430528-500MG
2-(4-Fluorophenoxy)-1-ethanamine 95%
2-(4-Fluorophenoxy)-1-ethanamine
Formel: C₈H₁₀FNO
Molecular Weight: 155,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00235183
CAS-Nummer: 6096-89-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR