2-Amino-6-fluoro-3-nitrobenzoic acid ethyl ester

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-amino-6-fluoro-3-nitrobenzoate

PC430529-500MG
APOSPC430529-500MGEA 210 EUR
APOSPC430529-500MG
2-Amino-6-fluoro-3-nitrobenzoic acid ethyl ester
2-Amino-6-fluoro-3-nitrobenzoic acid ethyl ester
Formel: C₉H₉FN₂O₄
Molecular Weight: 228,177 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00798156
CAS-Nummer: 150368-37-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR