2-(4-Fluoro-phenoxy)-acetamidine

Lieferant: Apollo Scientific

PC430530-250MG
APOSPC430530-250MGEA 232 EUR
APOSPC430530-250MG
2-(4-Fluoro-phenoxy)-acetamidine
2-(4-Fluoro-phenoxy)-acetamidine
Formel: C₈H₉FN₂O
Molecular Weight: 168,168 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07371514
CAS-Nummer: 284029-62-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR