3-(Aminomethyl)-5-fluoro-1,3-dihydro-2H-indol-2-one

Lieferant: Apollo Scientific

PC430531-250MG
APOSPC430531-250MGEA 232 EUR
APOSPC430531-250MG
3-(Aminomethyl)-5-fluoro-1,3-dihydro-2H-indol-2-one
3-(Aminomethyl)-5-fluoro-1,3-dihydro-2H-indol-2-one
Formel: C₉H₉FN₂O
Molecular Weight: 180,179 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06738904
CAS-Nummer: 518066-41-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR