4-(Trifluoromethyl)-6-(furan-2-yl)pyrimidine-2-thiol

Lieferant: Apollo Scientific

PC430533-100MG
APOSPC430533-100MGEA 252 EUR
APOSPC430533-100MG
4-(Trifluoromethyl)-6-(furan-2-yl)pyrimidine-2-thiol
4-(Trifluoromethyl)-6-(furan-2-yl)pyrimidine-2-thiol
Formel: C₉H₅F₃N₂OS
Molecular Weight: 246,209 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01929341
CAS-Nummer: 329710-06-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR