2-Chloro-4-fluoro-benzamidine

Lieferant: Apollo Scientific

PC430535-250MG
APOSPC430535-250MGEA 252 EUR
APOSPC430535-250MG
2-Chloro-4-fluoro-benzamidine
2-Chloro-4-fluoro-benzamidine
Formel: C₇H₆ClFN₂
Molecular Weight: 172,587 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05662904
CAS-Nummer: 582306-90-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR