2-Hydroxy-5-(4'-fluorophenyl)benzaldehyde

Lieferant: Apollo Scientific

Synonyme: 4-(4-Fluorophenyl)-2-formylphenol

PC430536-500MG
APOSPC430536-500MGEA 252 EUR
APOSPC430536-500MG
2-Hydroxy-5-(4'-fluorophenyl)benzaldehyde
2-Hydroxy-5-(4'-fluorophenyl)benzaldehyde
Formel: C₁₃H₉FO₂
Molecular Weight: 216,208 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06802113
CAS-Nummer: 847754-82-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR