N-(4-Fluorobenzyl)-6-chloropyrimidin-4-amine

Lieferant: Apollo Scientific

PC430537-500MG
APOSPC430537-500MGEA 274 EUR
APOSPC430537-500MG
N-(4-Fluorobenzyl)-6-chloropyrimidin-4-amine
N-(4-Fluorobenzyl)-6-chloropyrimidin-4-amine
Formel: C₁₁H₉ClFN₃
Molecular Weight: 237,661 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09756972
CAS-Nummer: 945896-79-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR