7-Fluoro-1,4-dihydro-6-methoxy-4-oxoquinoline-3-carboxylic acid

Lieferant: Apollo Scientific

PC430540-100MG
APOSPC430540-100MGEA 337 EUR
APOSPC430540-100MG
7-Fluoro-1,4-dihydro-6-methoxy-4-oxoquinoline-3-carboxylic acid
7-Fluoro-1,4-dihydro-6-methoxy-4-oxoquinoline-3-carboxylic acid
Formel: C₁₁H₈FNO₄
Molecular Weight: 237,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18157741
CAS-Nummer: 622369-36-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR