N-(4-Fluorobenzyl)-6-chloropyridazin-3-amine
Lieferant: Apollo Scientific
PC430543-500MG
APOSPC430543-500MGEA
337
EUR
APOSPC430543-500MG
N-(4-Fluorobenzyl)-6-chloropyridazin-3-amine
N-(4-Fluorobenzyl)-6-chloropyridazin-3-amine
Formel:
C₁₁H₉ClFN₃ Molecular Weight: 237,661 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07681959 CAS-Nummer: 872102-12-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...