tert-Butyl 4-(2-fluoro-6-hydroxyphenyl)piperidine-1-carboxylate

Lieferant: Apollo Scientific

PC430544-100MG
APOSPC430544-100MGEA 346 EUR
APOSPC430544-100MG
tert-Butyl 4-(2-fluoro-6-hydroxyphenyl)piperidine-1-carboxylate
tert-Butyl 4-(2-fluoro-6-hydroxyphenyl)piperidine-1-carboxylate
Formel: C₁₆H₂₂FNO₃
Molecular Weight: 295,349 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585455
CAS-Nummer: 1206970-27-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR