Diethyl 2-((3-cyano-4-fluorophenylamino)methylene)malonate
Lieferant: Apollo Scientific
PC430545-1G
APOSPC430545-1GEA
346
EUR
APOSPC430545-1G
Diethyl 2-((3-cyano-4-fluorophenylamino)methylene)malonate
Diethyl 2-((3-cyano-4-fluorophenylamino)methylene)malonate
Formel:
C₁₅H₁₅FN₂O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00277699 CAS-Nummer: 134811-49-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...