4-Fluoro-3-[5-(morpholin-4-ylmethyl)-1,2,4-oxadiazol-3-yl]benzaldehyde

Lieferant: Apollo Scientific

PC430546-250MG
APOSPC430546-250MGEA 346 EUR
APOSPC430546-250MG
4-Fluoro-3-[5-(morpholin-4-ylmethyl)-1,2,4-oxadiazol-3-yl]benzaldehyde
4-Fluoro-3-[5-(morpholin-4-ylmethyl)-1,2,4-oxadiazol-3-yl]benzaldehyde
Formel: C₁₄H₁₄FN₃O₃
Molecular Weight: 291.28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12026826
CAS-Nummer: 1119450-78-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR