5-Fluoroquinazolin-2-amine

Lieferant: Apollo Scientific

PC430547-500MG
APOSPC430547-500MGEA 346 EUR
APOSPC430547-500MG
5-Fluoroquinazolin-2-amine
5-Fluoroquinazolin-2-amine
Formel: C₈H₆FN₃
Molecular Weight: 163,152 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09998881
CAS-Nummer: 190273-81-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR