4,5-Difluoroanthranlic acid
Lieferant: Apollo Scientific
PC430548-250MG
APOSPC430548-250MGEA
367
EUR
APOSPC430548-250MG
4,5-Difluoroanthranlic acid
4,5-Difluoroanthranlic acid
Formel:
C₁₅H₈F₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD18157739 CAS-Nummer: 1258652-67-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...