(3,9-Diaza-spiro[5.5]undec-3-yl)(2-fluoro-phenyl)methanone
Lieferant: Apollo Scientific
PC430549-100MG
APOSPC430549-100MGEA
379
EUR
APOSPC430549-100MG
(3,9-Diaza-spiro[5.5]undec-3-yl)(2-fluoro-phenyl)methanone
(3,9-Diaza-spiro[5.5]undec-3-yl)(2-fluoro-phenyl)methanone
Formel:
C₁₆H₂₁FN₂O Molecular Weight: 276,349 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14585279 CAS-Nummer: 1206970-65-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...