7-Chloro-3-(3-fluorophenyl)naphthalene-1-carboxylic acid

Lieferant: Apollo Scientific

PC430550-250MG
APOSPC430550-250MGEA 389 EUR
APOSPC430550-250MG
7-Chloro-3-(3-fluorophenyl)naphthalene-1-carboxylic acid
7-Chloro-3-(3-fluorophenyl)naphthalene-1-carboxylic acid
Formel: C₁₇H₁₀ClFO₂
Molecular Weight: 300.712 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585483
CAS-Nummer: 1206969-68-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR