5-(4-(1,3-Dioxan-2-yl)-3-fluorophenyl)-2-chloropyridine
Lieferant: Apollo Scientific
PC430551-250MG
APOSPC430551-250MGEA
389
EUR
APOSPC430551-250MG
5-(4-(1,3-Dioxan-2-yl)-3-fluorophenyl)-2-chloropyridine
5-(4-(1,3-Dioxan-2-yl)-3-fluorophenyl)-2-chloropyridine
Formel:
C₁₅H₁₃ClFNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD14585365 CAS-Nummer: 1206969-79-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...