2-(4-Fluorophenyl)-2-(7-oxo-5H-pyrrolo[3,4-b]pyridin-6(7h)-yl)acetic acid

Lieferant: Apollo Scientific

PC430553-250MG
APOSPC430553-250MGEA 441 EUR
APOSPC430553-250MG
2-(4-Fluorophenyl)-2-(7-oxo-5H-pyrrolo[3,4-b]pyridin-6(7h)-yl)acetic acid
2-(4-Fluorophenyl)-2-(7-oxo-5H-pyrrolo[3,4-b]pyridin-6(7h)-yl)acetic acid
Formel: C₁₅H₁₁FN₂O₃
Molecular Weight: 286,258 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585256
CAS-Nummer: 1218657-27-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR