2-Fluoro-5-(6-Fluoropyridin-3-yl)benzaldehyde

Lieferant: Apollo Scientific

PC430554-500MG
APOSPC430554-500MGEA 441 EUR
APOSPC430554-500MG
2-Fluoro-5-(6-Fluoropyridin-3-yl)benzaldehyde
2-Fluoro-5-(6-Fluoropyridin-3-yl)benzaldehyde
Formel: C₁₂H₇F₂NO
Molecular Weight: 219.187 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585404
CAS-Nummer: 1206969-49-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR