Diethyl 2-((4-fluorophenylamino)methylene)malonate
Lieferant: Apollo Scientific
PC430555-1G
APOSPC430555-1GEA
461
EUR
APOSPC430555-1G
Diethyl 2-((4-fluorophenylamino)methylene)malonate
Diethyl 2-((4-fluorophenylamino)methylene)malonate
Formel:
C₁₄H₁₆FNO₄ Molecular Weight: 281,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00111036 CAS-Nummer: 26832-96-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...