(S)-Methyl 3-(4-(2-fluorobenzyloxy)phenyl)-2-aminopropanoate
Lieferant: Apollo Scientific
PC430556-250MG
APOSPC430556-250MGEA
461
EUR
APOSPC430556-250MG
(S)-Methyl 3-(4-(2-fluorobenzyloxy)phenyl)-2-aminopropanoate
(S)-Methyl 3-(4-(2-fluorobenzyloxy)phenyl)-2-aminopropanoate
Formel:
C₁₇H₁₈FNO₃ Molecular Weight: 303,328 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14570123 CAS-Nummer: 1212808-41-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...