5-(2-Chloro-5-(trifluoromethyl)phenoxy)pyrazin-2-amine

Lieferant: Apollo Scientific

PC430557-250MG
APOSPC430557-250MGEA 461 EUR
APOSPC430557-250MG
5-(2-Chloro-5-(trifluoromethyl)phenoxy)pyrazin-2-amine
5-(2-Chloro-5-(trifluoromethyl)phenoxy)pyrazin-2-amine
Formel: C₁₁H₇ClF₃N₃O
Molecular Weight: 289,641 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585379
CAS-Nummer: 1206970-21-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR