7-Chloro-n-(2-fluorophenyl)thiazolo[5,4-d]pyrimidin-2-amine

Lieferant: Apollo Scientific

PC430559-250MG
APOSPC430559-250MGEA 483 EUR
APOSPC430559-250MG
7-Chloro-n-(2-fluorophenyl)thiazolo[5,4-d]pyrimidin-2-amine
7-Chloro-n-(2-fluorophenyl)thiazolo[5,4-d]pyrimidin-2-amine
Formel: C₁₁H₆ClFN₄S
Molecular Weight: 280,709 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16987529
CAS-Nummer: 1242240-95-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR