6-Chloro-n-(4-fluorophenyl)pyridin-3-amine

Lieferant: Apollo Scientific

PC430560-250MG
APOSPC430560-250MGEA 483 EUR
APOSPC430560-250MG
6-Chloro-n-(4-fluorophenyl)pyridin-3-amine
6-Chloro-n-(4-fluorophenyl)pyridin-3-amine
Formel: C₁₁H₈ClFN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD16987532
CAS-Nummer: 509953-60-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR