2-(Chloromethyl)-4-(trifluoromethyl)-6-(naphthalen-3-yl)pyrimidine

Lieferant: Apollo Scientific

PC430562-250MG
APOSPC430562-250MGEA 483 EUR
APOSPC430562-250MG
2-(Chloromethyl)-4-(trifluoromethyl)-6-(naphthalen-3-yl)pyrimidine
2-(Chloromethyl)-4-(trifluoromethyl)-6-(naphthalen-3-yl)pyrimidine
Formel: C₁₆H₁₀ClF₃N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD14585229
CAS-Nummer: 1206970-47-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR