(2,8-Diaza-spiro[4.5]dec-2-yl)(2-fluoro-phenyl)methanone
Lieferant: Apollo Scientific
PC430563-250MG
APOSPC430563-250MGEA
483
EUR
APOSPC430563-250MG
(2,8-Diaza-spiro[4.5]dec-2-yl)(2-fluoro-phenyl)methanone
(2,8-Diaza-spiro[4.5]dec-2-yl)(2-fluoro-phenyl)methanone
Formel:
C₁₅H₁₉FN₂O Molecular Weight: 262,323 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14585327 CAS-Nummer: 1206969-90-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...