8'-Fluoro-5'-hydroxy-spiro[cyclohexane-1,4'(1'h)-quinazolin]-2'(3'h)-one

Lieferant: Apollo Scientific

PC430564-250MG
APOSPC430564-250MGEA 483 EUR
APOSPC430564-250MG
8'-Fluoro-5'-hydroxy-spiro[cyclohexane-1,4'(1'h)-quinazolin]-2'(3'h)-one
8'-Fluoro-5'-hydroxy-spiro[cyclohexane-1,4'(1'h)-quinazolin]-2'(3'h)-one
Formel: C₁₃H₁₅FN₂O₂
Molecular Weight: 250,269 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585364
CAS-Nummer: 939768-67-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR