1-(2-Bromo-4-fluorobenzyl)-6,7-dihydro-1H-pyrrolo[3,2-c]pyridin-4(5H)-one

Lieferant: Apollo Scientific

PC430565-500MG
APOSPC430565-500MGEA 483 EUR
APOSPC430565-500MG
1-(2-Bromo-4-fluorobenzyl)-6,7-dihydro-1H-pyrrolo[3,2-c]pyridin-4(5H)-one
1-(2-Bromo-4-fluorobenzyl)-6,7-dihydro-1H-pyrrolo[3,2-c]pyridin-4(5H)-one
Formel: C₁₄H₁₂BrFN₂O
Molecular Weight: 323,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585354
CAS-Nummer: 1206969-77-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR