N-(4-Fluorobenzyl)-6-chloropyrazin-2-amine

Lieferant: Apollo Scientific

PC430566-250MG
APOSPC430566-250MGEA 504 EUR
APOSPC430566-250MG
N-(4-Fluorobenzyl)-6-chloropyrazin-2-amine
N-(4-Fluorobenzyl)-6-chloropyrazin-2-amine
Formel: C₁₁H₉ClFN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11790282
CAS-Nummer: 629670-16-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR