2-(3-Fluorophenoxy)-6-chloropyrazine
Lieferant: Apollo Scientific
PC430567-250MG
APOSPC430567-250MGEA
504
EUR
APOSPC430567-250MG
2-(3-Fluorophenoxy)-6-chloropyrazine
2-(3-Fluorophenoxy)-6-chloropyrazine
Formel:
C₁₀H₆ClFN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD09952090 CAS-Nummer: 894416-95-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...