4-(3-Fluorophenoxy)-2-chloropyrimidine

Lieferant: Apollo Scientific

PC430568-250MG
APOSPC430568-250MGEA 504 EUR
APOSPC430568-250MG
4-(3-Fluorophenoxy)-2-chloropyrimidine
4-(3-Fluorophenoxy)-2-chloropyrimidine
Formel: C₁₀H₆ClFN₂O
Molecular Weight: 224,619 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16712408
CAS-Nummer: 1242240-84-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR