Ethyl 4-chloro-7-fluoro-6-methoxyquinoline-3-carboxylate
Lieferant: Apollo Scientific
PC430569-250MG
APOSPC430569-250MGEA
504
EUR
APOSPC430569-250MG
Ethyl 4-chloro-7-fluoro-6-methoxyquinoline-3-carboxylate
Ethyl 4-chloro-7-fluoro-6-methoxyquinoline-3-carboxylate
Formel:
C₁₃H₁₁ClFNO₃ Molecular Weight: 283,683 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16036879 CAS-Nummer: 311346-71-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...