7-(Trifluoromethyl)benzo[b]thiophene-2-carbaldehyde

Lieferant: Apollo Scientific
Achtung

PC430570-250MG PC430570-1G
APOSPC430570-250MGEA 0 EUR
APOSPC430570-250MG APOSPC430570-1G
7-(Trifluoromethyl)benzo[b]thiophene-2-carbaldehyde
7-(Trifluoromethyl)benzo[b]thiophene-2-carbaldehyde
Formel: C₁₀H₅F₃OS
Lagertemperatur: Gefrierschrank
MDL: MFCD20926386
CAS-Nummer: 1353011-97-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR