3-(6-Fluoro-1-oxoisoindolin-2-yl)propanoic acid
Lieferant: Apollo Scientific
PC430571-250MG
APOSPC430571-250MGEA
504
EUR
APOSPC430571-250MG
3-(6-Fluoro-1-oxoisoindolin-2-yl)propanoic acid
3-(6-Fluoro-1-oxoisoindolin-2-yl)propanoic acid
Formel:
C₁₁H₁₀FNO₃ Molecular Weight: 223,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14585265 CAS-Nummer: 1206970-40-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...