3-(3,4-Difluorophenyl)-3-oxo-propionic acid ethyl ester

Lieferant: Apollo Scientific

Synonyme: Ethyl 3-(3,4-difluorophenyl)-3-oxopropanoate

PC430572-1G
APOSPC430572-1GEA 112 EUR
APOSPC430572-1G
3-(3,4-Difluorophenyl)-3-oxo-propionic acid ethyl ester
3-(3,4-Difluorophenyl)-3-oxo-propionic acid ethyl ester
Formel: C₁₁H₁₀F₂O₃
Molecular Weight: 228,192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03414546
CAS-Nummer: 252955-07-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR