4-(3-Fluorophenoxy)-6-chloropyrimidine

Lieferant: Apollo Scientific

PC430573-250MG
APOSPC430573-250MGEA 513 EUR
APOSPC430573-250MG
4-(3-Fluorophenoxy)-6-chloropyrimidine
4-(3-Fluorophenoxy)-6-chloropyrimidine
Formel: C₁₀H₆ClFN₂O
Molecular Weight: 224,619 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10456892
CAS-Nummer: 1017067-02-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR