N-Trifluoroacetyl-l-phenylalanine

Lieferant: Apollo Scientific
Achtung

PC430575-1G
APOSPC430575-1GEA 524 EUR
APOSPC430575-1G
N-Trifluoroacetyl-l-phenylalanine
N-Trifluoroacetyl-l-phenylalanine
Formel: C₁₁H₁₀F₃NO₃
Molecular Weight: 261,197 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00800548
CAS-Nummer: 350-09-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR