Perfluorophenyl 4-fluorobenzoate

Lieferant: Apollo Scientific

PC430576-1G
APOSPC430576-1GEA 524 EUR
APOSPC430576-1G
Perfluorophenyl 4-fluorobenzoate
Perfluorophenyl 4-fluorobenzoate
Formel: C₁₃H₄F₆O₂
Molecular Weight: 306,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585474
CAS-Nummer: 193947-55-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR