2-(6-Fluoro-1-oxoisoindolin-2-yl)acetic acid
Lieferant: Apollo Scientific
PC430577-250MG
APOSPC430577-250MGEA
535
EUR
APOSPC430577-250MG
2-(6-Fluoro-1-oxoisoindolin-2-yl)acetic acid
2-(6-Fluoro-1-oxoisoindolin-2-yl)acetic acid
Formel:
C₁₀H₈FNO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD14585261 CAS-Nummer: 1206969-83-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...