Ethyl 6-(4-(6-(3-fluorophenyl)pyridine-3-carboxamido)piperidin-1-yl)pyridazine-3-carboxylate

Lieferant: Apollo Scientific

PC430578-250MG
APOSPC430578-250MGEA 546 EUR
APOSPC430578-250MG
Ethyl 6-(4-(6-(3-fluorophenyl)pyridine-3-carboxamido)piperidin-1-yl)pyridazine-3-carboxylate
Ethyl 6-(4-(6-(3-fluorophenyl)pyridine-3-carboxamido)piperidin-1-yl)pyridazine-3-carboxylate
Formel: C₂₄H₂₄FN₅O₃
Molecular Weight: 449,477 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22574828
CAS-Nummer: 1393330-39-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR