5-Bromo-4-fluoro-2-hydroxybenzaldehyde 98%

Lieferant: Apollo Scientific
Achtung

PC430579-5G
APOSPC430579-5GEA 168 EUR
APOSPC430579-5G
5-Bromo-4-fluoro-2-hydroxybenzaldehyde 98%
5-Bromo-4-fluoro-2-hydroxybenzaldehyde
Formel: C₇H₄BrFO₂
Molecular Weight: 219.008 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06739493
CAS-Nummer: 399-00-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR