4,4,4-Trifluoro-1-(3-oxo-3,4-dihydro-2H-benzo[1,4]oxazin-6-yl)-butane-1,3-dione

Lieferant: Apollo Scientific

PC430580-250MG
APOSPC430580-250MGEA 546 EUR
APOSPC430580-250MG
4,4,4-Trifluoro-1-(3-oxo-3,4-dihydro-2H-benzo[1,4]oxazin-6-yl)-butane-1,3-dione
4,4,4-Trifluoro-1-(3-oxo-3,4-dihydro-2H-benzo[1,4]oxazin-6-yl)-butane-1,3-dione
Formel: C₁₂H₈F₃NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD14585419
CAS-Nummer: 943994-23-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR