6-Fluoro-2-(pyrrolidin-3-yl)isoindolin-1-one

Lieferant: Apollo Scientific

PC430581-250MG
APOSPC430581-250MGEA 556 EUR
APOSPC430581-250MG
6-Fluoro-2-(pyrrolidin-3-yl)isoindolin-1-one
6-Fluoro-2-(pyrrolidin-3-yl)isoindolin-1-one
Formel: C₁₂H₁₃FN₂O
Molecular Weight: 220,243 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585250
CAS-Nummer: 1206969-88-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR