3-(2-Fluorophenyl)-[1,2,4]triazolo[4,3-a]pyridine-8-carboxylic acid
Lieferant: Apollo Scientific
PC430582-250MG
APOSPC430582-250MGEA
556
EUR
APOSPC430582-250MG
3-(2-Fluorophenyl)-[1,2,4]triazolo[4,3-a]pyridine-8-carboxylic acid
3-(2-Fluorophenyl)-[1,2,4]triazolo[4,3-a]pyridine-8-carboxylic acid
Formel:
C₁₃H₈FN₃O₂ Molecular Weight: 257,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07376996 CAS-Nummer: 1206970-37-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...