4-(2-(2-(Trifluoromethyl)-4-nitrophenoxy)ethyl)morpholine

Lieferant: Apollo Scientific

PC430585-500MG
APOSPC430585-500MGEA 567 EUR
APOSPC430585-500MG
4-(2-(2-(Trifluoromethyl)-4-nitrophenoxy)ethyl)morpholine
4-(2-(2-(Trifluoromethyl)-4-nitrophenoxy)ethyl)morpholine
Formel: C₁₃H₁₅F₃N₂O₄
Molecular Weight: 320,264 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585384
CAS-Nummer: 850797-92-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR