(5-Chloro-2-iodophenyl)(2-fluoro-6-methoxyphenyl)methanone

Lieferant: Apollo Scientific

PC430586-1G
APOSPC430586-1GEA 576 EUR
APOSPC430586-1G
(5-Chloro-2-iodophenyl)(2-fluoro-6-methoxyphenyl)methanone
(5-Chloro-2-iodophenyl)(2-fluoro-6-methoxyphenyl)methanone
Formel: C₁₄H₉ClFIO₂
Molecular Weight: 390,576 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11520937
CAS-Nummer: 1233025-91-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR