2,2,3-Trifluoro-2,3-dihydrobenzo[1,4]-dioxin-5-ol

Lieferant: Apollo Scientific

PC430587-500MG
APOSPC430587-500MGEA 576 EUR
APOSPC430587-500MG
2,2,3-Trifluoro-2,3-dihydrobenzo[1,4]-dioxin-5-ol
2,2,3-Trifluoro-2,3-dihydrobenzo[1,4]-dioxin-5-ol
Formel: C₈H₅F₃O₃
Molecular Weight: 206,119 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04972979
CAS-Nummer: 1008452-11-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR